Anti-ENDOG Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513513
CAPIPA5-13513EA 2561.45 CAD
CAPIPA5-13513
Anti-ENDOG Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine based on sequence homology. ENDOG is a nuclear encoded endonuclease that is localized in the mitochondrion. The protein is widely distributed among animals and cleaves DNA at GC tracts. This protein is capable of generating the RNA primers required by DNA polymerase gamma to initiate replication of mitochondrial DNA.

Recommended Dilutions: Western Blot: 1:1000, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: ENDOG (endonuclease G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR