Anti-FKBP14 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513514
CAPIPA5-13514EA 2561.45 CAD
CAPIPA5-13514
Anti-FKBP14 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse based on sequence homology. PPIases accelerate the folding of proteins during protein synthesis.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: FKBP14 (FK506 binding protein 14, 22 kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR