Anti-GNS Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513517
CAPIPA5-13517EA 2561.45 CAD
CAPIPA5-13517
Anti-GNS Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine and mouse based on sequence homology. GNS is a lysosomal enzyme found in all cells. It is involved in the catabolism of heparin, heparan sulphate, and keratan sulphate. Deficiency of this enzyme results in the accumulation of undegraded substrate and the lysosomal storage disorder mucopolysaccharidosis type IIID (Sanfilippo D syndrome). Mucopolysaccharidosis type IIID is the least common of the four subtypes of Sanfilippo syndrome.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: GNS (glucosamine (N-acetyl)-6-sulfatase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR