Anti-GZMB Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513518
CAPIPA5-13518EA 2801.79 CAD
CAPIPA5-13518
Anti-GZMB Rabbit Polyclonal Antibody
Antibodies
Cytolytic T lymphocytes (CTL) and natural killer (NK) cells share the remarkable ability to recognize, bind, and lyse specific target cells. They are thought to protect their host by lysing cells bearing on their surface 'nonself' antigens, usually peptides or proteins resulting from infection by intracellular pathogens. The protein is crucial for the rapid induction of target cell apoptosis by CTL in cell-mediated immune response.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: GZMB (granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR