Anti-CNN3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513524
CAPIPA5-13524EA 2561.45 CAD
CAPIPA5-13524
Anti-CNN3 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine based on sequence homology. Calponin-3 is a protein with a markedly acidic C terminus; the basic N-terminus is highly homologous to the N-terminus of a related gene, CNN1. This protein is associated with the cytoskeleton but is not involved in contraction.

Flow Cytometry: 1:10-1:50, Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: CNN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR