Anti-c-Fos Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513531
CAPIPA5-13531EA 2561.45 CAD
CAPIPA5-13531
Anti-c-Fos Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine and rat based on sequence homology. The Fos family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These leucine zipper proteins can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. In some cases, expression of the FOS gene has also been associated with apoptotic cell death.

Flow Cytometry: 1:10-1:50, Western Blot: 1:1000

Type: Primary
Antigen: c-Fos
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR