Anti-LMNB1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513532
CAPIPA5-13532EA 2801.79 CAD
CAPIPA5-13532
Anti-LMNB1 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. The nuclear lamina consists of a two-dimensional matrix of proteins located next to the inner nuclear membrane. The lamin family of proteins make up the matrix and are highly conserved in evolution. During mitosis, the lamina matrix is reversibly disassembled as the lamin proteins are phosphorylated. Lamin proteins are thought to be involved in nuclear stability, chromatin structure and gene expression. Vertebrate lamins consist of two types, A and B. Lamin B1 is one of the two B type proteins, B1.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: LMNB1 (lamin B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR