Anti-Decorin Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513538
CAPIPA5-13538EA 2801.79 CAD
CAPIPA5-13538
Anti-Decorin Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine and porcine based on sequence homology. DCN is a small cellular or pericellular matrix proteoglycan that is closely related in structure to biglycan protein. The protein and biglycan are thought to be the result of a gene duplication. This protein is a component of connective tissue, binds to type I collagen fibrils, and plays a role in matrix assembly. It contains one attached glycosaminoglycan chain. This protein is capable of suppressing the growth of various tumor cell lines.

Flow Cytometry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:10-1:50, Western Blot: 1:1000

Type: Primary
Antigen: Decorin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR