Anti-ARPC3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513549
CAPIPA5-13549EA 2561.45 CAD
CAPIPA5-13549
Anti-ARPC3 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine based on sequence homology. ARPC3 is one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein, the p21 subunit, has yet to be determined.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: ARPC3 (actin related protein 2/3 complex, subunit 3, 21kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR