Anti-CALD Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513559
CAPIPA5-13559EA 2561.45 CAD
CAPIPA5-13559
Anti-CALD Rabbit Polyclonal Antibody
CALD1 is a calmodulin- and actin-binding protein that plays an essential role in the regulation of smooth muscle and nonmuscle contraction. The conserved domain of this protein possesses the binding activities to Ca(2+)-calmodulin, actin, tropomyosin, myosin, and phospholipids. This protein is a potent inhibitor of the actin-tropomyosin activated myosin MgATPase, and serves as a mediating factor for Ca(2+)-dependent inhibition of smooth muscle contraction.

Flow Cytometry: 1:10-1:50, Immunohistochemistry: 1:10-1:50, Western Blot: 1:1000

Type: Primary
Antigen: CALD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR