Anti-CDCA3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513564
CAPIPA5-13564EA 2801.79 CAD
CAPIPA5-13564
Anti-CDCA3 Rabbit Polyclonal Antibody
Antibodies
CDCA3 is a F-box-like protein which is required for entry into mitosis. It acts by participating to E3 ligase complexes that mediate the ubiquitination and degradation of WEE1 kinase at G2/M phase.

Flow Cytometry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:50 - 1:100, Western Blot: 1:1000

Type: Primary
Antigen: CDCA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR