Anti-CFHR1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513565
CAPIPA5-13565EA 2561.45 CAD
CAPIPA5-13565
Anti-CFHR1 Rabbit Polyclonal Antibody
CFHL1 might be involved in complement regulation. The protein can associate with lipoproteins and may play a role in lipid metabolism.

Flow Cytometry: 1:10-1:50, Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: CFHR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR