Anti-RPS9 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513569
CAPIPA5-13569EA 2444.36 CAD
CAPIPA5-13569
Anti-RPS9 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPS9 is a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S4P family of ribosomal proteins. It is located in the cytoplasm.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry (Paraffin): 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: RPS9 (ribosomal protein S9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR