Anti-RRM2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513570
CAPIPA5-13570EA 2573.22 CAD
CAPIPA5-13570
Anti-RRM2 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with non-human primate based on sequence homology. RRM2 is one of two non-identical subunits for ribonucleotide reductase. This reductase catalyzes the formation of deoxyribonucleotides from ribonucleotides. Synthesis of the protein (M2) is regulated in a cell-cycle dependent fashion.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: RRM2 (ribonucleotide reductase M2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR