Anti-SRSF5 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513571
CAPIPA5-13571EA 2801.79 CAD
CAPIPA5-13571
Anti-SRSF5 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. SFRS5 plays a role in constitutive splicing and can modulate the selection of alternative splice sites.

Recommended Dilutions: Western Blot: 1:1000, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: SRSF5 (serine/arginine-rich splicing factor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR