Anti-KLF11 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513578
CAPIPA5-13578EA 2515.44 CAD
CAPIPA5-13578
Anti-KLF11 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse based on sequence homology. TIEG2 is a transcription factor. The protein activates the epsilon- and gamma-globin gene promoters and, to a much lower degree, the beta-globin gene and represses promoters containing SP1-like binding inhibiting cell growth. It represses transcription of SMAD7 which enhances TGF-beta signaling. It induces apoptosis.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: KLF11 (Kruppel-like factor 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR