Anti-AZGP1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513580
CAPIPA5-13580EA 2801.79 CAD
CAPIPA5-13580
Anti-AZGP1 Rabbit Polyclonal Antibody
Antibodies
AZGP1 stimulates lipid degradation in adipocytes and causes the extensive fat losses associated with some advanced cancers. It may bind polyunsaturated fatty acids.

Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: AZGP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR