Anti-FGF18 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513596
CAPIPA5-13596EA 2444.36 CAD
CAPIPA5-13596
Anti-FGF18 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. FGF18 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. It has been shown in vitro that this protein is able to induce neurite outgrowth in PC12 cells. Studies of the similar proteins in mouse and chick suggested that this protein is a pleiotropic growth factor that stimulates proliferation in a number of tissues, most notably the liver and small intestine. Knockout studies of the similar gene in mice implied the role of this protein in regulating proliferation and differentiation of midline cerebellar structures.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: FGF18 (fibroblast growth factor 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR