Anti-FOXO1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513597
CAPIPA5-13597EA 2573.22 CAD
CAPIPA5-13597
Anti-FOXO1 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine and porcine based on sequence homology. FKHR belongs to the forkhead family of transcription factors which are characterized by a distinct forkhead domain.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: FOXO1 (forkhead box O1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR