Anti-HTATSF1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513606
CAPIPA5-13606EA 2561.45 CAD
CAPIPA5-13606
Anti-HTATSF1 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse based on sequence homology. HTATSF1 functions as a cofactor for the stimulation of transcriptional elongation by HIV-1 Tat, which binds to the HIV-1 promoter through Tat-TAR interaction. This protein may also serve as a dual-function factor to couple transcription and splicing and to facilitate their reciprocal activation.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: HTATSF1 (HIV-1 Tat specific factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR