Anti-NUP155 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513615
CAPIPA5-13615EA 2561.45 CAD
CAPIPA5-13615
Anti-NUP155 Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse and rat based on sequence homology. Nucleoporins are the main components of the nuclear pore complex (NPC) of eukaryotic cells. They are involved in the bidirectional trafficking of molecules, especially mRNAs and proteins, between the nucleus and the cytoplasm. The protein does not contain the typical FG repeat sequences found in most vertebrate nucleoporins.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: NUP155 (nucleoporin 155kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR