Anti-SPCS3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513630
CAPIPA5-13630EA 2561.45 CAD
CAPIPA5-13630
Anti-SPCS3 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine based on sequence homology. SPCS3 is a component of the microsomal signal peptidase complex which removes signal peptides from nascent proteins as they are translocated into the lumen of the endoplasmic reticulum.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: SPCS3 (signal peptidase complex subunit 3 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR