Anti-AOX1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513659
CAPIPA5-13659EA 2573.22 CAD
CAPIPA5-13659
Anti-AOX1 Rabbit Polyclonal Antibody
Antibodies
AOX1 catalyzes: An aldehyde + H2O + O2 = a carboxylic acid + H2O2.

Flow Cytometry: 1:10-1:50, Western Blot: 1:1000

Type: Primary
Antigen: AOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR