Anti-CK10 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513663
CAPIPA5-13663EA 2573.22 CAD
CAPIPA5-13663
Anti-CK10 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with rat based on sequence homology. KRT10 is a member of the type I (acidic) cytokeratin family, which belongs to the superfamily of intermediate filament (IF) proteins. Keratins are heteropolymeric structural proteins which form the intermediate filament. These filaments, along with actin microfilaments and microtubules, compose the cytoskeleton of epithelial cells. Mutations in its gene are associated with epidermolytic hyperkeratosis.

Flow Cytometry: 1:10-1:50, Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: CK10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR