Anti-LRG1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513664
CAPIPA5-13664EA 2561.45 CAD
CAPIPA5-13664
Anti-LRG1 Rabbit Polyclonal Antibody
The leucine-rich repeat (LRR) family of proteins, including LRG1, have been shown to be involved in protein-protein interaction, signal transduction, and cell adhesion and development. LRG1 is expressed during granulocyte differentiation.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: LRG1 (leucine-rich alpha-2-glycoprotein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR