Anti-TIMP1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513668
CAPIPA5-13668EA 2353.26 CAD
CAPIPA5-13668
Anti-TIMP1 Rabbit Polyclonal Antibody
Antibodies
TIMP1 belongs to the TIMP family. The proteins in this family are natural inhibitors of the matrix metalloproteinases (MMPs), a group of peptidases involved in degradation of the extracellular matrix. In addition to its inhibitory role against most of the known MMPs, the protein is able to promote cell proliferation in a wide range of cell types, and may also have an anti-apoptotic function.

  • Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunohistochemistry: 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: TIMP1 (TIMP metallopeptidase inhibitor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR