Anti-CYP2C19 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513669
CAPIPA5-13669EA 2801.79 CAD
CAPIPA5-13669
Anti-CYP2C19 Rabbit Polyclonal Antibody
Antibodies
CYP2C19 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is known to metabolize many xenobiotics, including the anticonvulsive drug mephenytoin, omeprazole, diazepam and some barbiturates. Polymorphism within its gene is associated with variable ability to metabolize mephenytoin, known as the poor metabolizer and extensive metabolizer phenotypes.

Immunohistochemistry: 1:10-1:50, Western Blot: 1:1000

Type: Primary
Antigen: CYP2C19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR