Anti-SERPINA7 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513673
CAPIPA5-13673EA 2561.45 CAD
CAPIPA5-13673
Anti-SERPINA7 Rabbit Polyclonal Antibody
SERPINA7 is a major thyroid hormone transport protein in serum.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: SERPINA7 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR