Anti-MAML3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513678
CAPIPA5-13678EA 2286.72 CAD
CAPIPA5-13678
Anti-MAML3 Rabbit Polyclonal Antibody
Antibodies
It has been shown to amplify NOTCH-induced transcription of HES1.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: MAML3 (mastermind-like 3 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR