Anti-PRAME Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513679
CAPIPA5-13679EA 2801.79 CAD
CAPIPA5-13679
Anti-PRAME Rabbit Polyclonal Antibody
Antibodies
MAPE is an antigen that is predominantly expressed in human melanomas and that is recognized by cytolytic T lymphocytes. It is not expressed in normal tissues, except testis. This expression pattern is similar to that of other CT antigens, such as MAGE, BAGE and GAGE. However, unlike these other CT antigens, its gene is also expressed in acute leukemias.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: PRAME (preferentially expressed antigen in melanoma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR