Anti-FBP2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513689
CAPIPA5-13689EA 2561.45 CAD
CAPIPA5-13689
Anti-FBP2 Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine and rabbit based on sequence homology. FBP2 is a gluconeogenesis regulatory enzyme which catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: FBP2 (fructose-1,6-bisphosphatase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR