Anti-TOMM70A Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513705
CAPIPA5-13705EA 2444.36 CAD
CAPIPA5-13705
Anti-TOMM70A Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse and rat based on sequence homology. The translocase of outer mitochondrial membrane (TOM) complex is a multisubunit complex involved in the recognition, unfolding, and translocation of preproteins into the mitochondria.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: TOMM70A (translocase of outer mitochondrial membrane 70 homolog A (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR