Anti-MTMR8 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513714
CAPIPA5-13714EA 2561.45 CAD
CAPIPA5-13714
Anti-MTMR8 Rabbit Polyclonal Antibody
MTMR8 is a myotubularin-related protein that is atypical to most other members of the myotubularin-related protein family because it has no dual-specificity phosphatase domain. The encoded protein contains a double-helical motif similar to the SET interaction domain, which is thought to have a role in the control of cell proliferation. In mouse, a protein similar to the encoded protein binds with MTMR7, and together they dephosphorylate phosphatidylinositol 3-phosphate and inositol 1,3-bisphosphate.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50

Type: Primary
Antigen: MTMR8 (myotubularin related protein 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR