Anti-GRK6 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513730
CAPIPA5-13730EA 2444.36 CAD
CAPIPA5-13730
Anti-GRK6 Rabbit Polyclonal Antibody
This antibody is predicted to react with rat based on sequence homology. GRK6 is a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating their deactivation.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: GRK6 (G protein-coupled receptor kinase 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR