Anti-RIOK2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513732
CAPIPA5-13732EA 2561.45 CAD
CAPIPA5-13732
Anti-RIOK2 Rabbit Polyclonal Antibody
RIOK2 belongs to the protein kinase superfamily; RIO-type Ser/Thr kinase family. Serine/threonine protein kinases, such as RIOK2, phosphorylate the OH group of serine or threonine (which have similar sidechains). Activity of these protein kinases can be regulated by specific events (e.g. DNA damage), as well as numerous chemical signals, including cAMP/cGMP, Diacylglycerol, and Ca2+/calmodulin.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RIOK2 (RIO kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR