Anti-TAOK3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513735
CAPIPA5-13735EA 2444.36 CAD
CAPIPA5-13735
Anti-TAOK3 Rabbit Polyclonal Antibody
This antibody is predicted to react with chicken, mouse, rat and Xenopus based on sequence homology. TAOK3 is negatively regulated by epidermal growth factor (EGF) and Inhibits the basal activity of Jun kinase. When TAOK3 is overexpressed, it may activate ERK1/ERK2 and JNK/SAPK.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50

Type: Primary
Antigen: TAOK3 (TAO kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR