Anti-PXK Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513743
CAPIPA5-13743EA 2561.45 CAD
CAPIPA5-13743
Anti-PXK Rabbit Polyclonal Antibody
This antibody is predicted to react with mouse and rat based on sequence homology. PXK is widely expressed in all tissues examined except in heart. It binds to and modulates brain Na,K-ATPase subunits ATP1B1 and ATP1B3 and may thereby participate in the regulation of electrical excitability and synaptic transmission. However, the protein kinase domain is predicted to be catalytically inactive.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PXK (PX domain containing serine/threonine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR