Anti-PRKCD Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513744
CAPIPA5-13744EA 2572.49 CAD
CAPIPA5-13744
Anti-PRKCD Rabbit Polyclonal Antibody
Antibodies
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells. PKC delta is one of the PKC family members, and may play a role in antigen-dependent control of B-cell function.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: PRKCD (protein kinase C, delta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR