Anti-SCYL2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513751
CAPIPA5-13751EA 2561.45 CAD
CAPIPA5-13751
Anti-SCYL2 Rabbit Polyclonal Antibody
SCYL2 is a component of AP2-containing clathrin coated structures at the plasma membrane or of endocytic coated vesicles. SCYL2 is a probable serine/threonine-protein kinase that phosphorylates, in vitro, the beta2-subunit of the plasma membrane adapter complex AP2 and other proteins in presence of poly-L-lysine. SCYL2 may also regulate clathrin-dependent trafficking between the TGN and/or the endosomal system.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:10-1:50

Type: Primary
Antigen: SCYL2 (SCY1-like 2 (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR