Anti-AKT2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513761
CAPIPA5-13761EA 2801.79 CAD
CAPIPA5-13761
Anti-AKT2 Rabbit Polyclonal Antibody
Antibodies
AKT2 is a putative oncogenic protein belonging to a subfamily of serine/threonine kinases containing SH2-like (Src homology 2-like) domains. Furthermore, AKT2 was shown to be amplified and overexpressed in 2 of 8 ovarian carcinoma cell lines and 2 of 15 primary ovarian tumors. Overexpression of AKT2 contributes to the malignant phenotype of a subset of human ductal pancreatic cancers. AKT2 is a general protein kinase capable of phophorylating several known proteins.

Flow Cytometry: 1:10-1:50, Immunocytochemistry: 1:10-1:50, Immunofluorescence: 1:10-1:50, Immunohistochemistry: 1:10-1:50, Western Blot: 1:1000

Type: Primary
Antigen: AKT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR