Anti-MOB1B Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513769
CAPIPA5-13769EA 2573.22 CAD
CAPIPA5-13769
Anti-MOB1B Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. MOB4A is similar to the yeast Mob1 protein. Yeast Mob1 binds Mps1p, a protein kinase essential for spindle pole body duplication and mitotic checkpoint regulation. Mob4A regulates NDR1 and NDR2 kinase activity, widely expressed nuclear serine-threonine kinases that have been implicated in cell proliferation and/or tumor progression.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunohistochemistry: 1:10-1:50

Type: Primary
Antigen: MOB1B (MOB kinase activator 1B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR