Anti-PDK2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513774
CAPIPA5-13774EA 2286.72 CAD
CAPIPA5-13774
Anti-PDK2 Rabbit Polyclonal Antibody
Antibodies
PDK2 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.

Recommended Dilutions: Western Blot: 1:100-1:500, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: PDK2 (pyruvate dehydrogenase kinase, isozyme 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR