Anti-PDK4 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513778
CAPIPA5-13778EA 2444.36 CAD
CAPIPA5-13778
Anti-PDK4 Rabbit Polyclonal Antibody
PDK4 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunohistochemistry: 1:10-1:50

Type: Primary
Antigen: PDK4 (pyruvate dehydrogenase kinase, isozyme 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR