Anti-PKLR Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513787
CAPIPA5-13787EA 2444.36 CAD
CAPIPA5-13787
Anti-PKLR Rabbit Polyclonal Antibody
PKLR is a pyruvate kinase that catalyzes the production of phosphoenolpyruvate from pyruvate and ATP. Defects in this enzyme, due to gene mutations or genetic variations, are the common cause of chronic hereditary nonspherocytic hemolytic anemia (CNSHA or HNSHA).

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: PKLR (pyruvate kinase, liver and RBC)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR