Anti-Pyruvate Kinase Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513789
CAPIPA5-13789EA 2561.45 CAD
CAPIPA5-13789
Anti-Pyruvate Kinase Rabbit Polyclonal Antibody
This antibody is predicted to react with rat and rabbit based on sequence homology. PKM2 is a pyruvate kinase that catalyzes the production of phosphoenolpyruvate from pyruvate and ATP. This protein has been shown to interact with thyroid hormone, and thus may mediate cellular metabolic effects induced by thyroid hormones. This protein has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: Pyruvate Kinase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR