Anti-MKNK2 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513809
CAPIPA5-13809EA 2801.79 CAD
CAPIPA5-13809
Anti-MKNK2 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with mouse and rat based on sequence homology. MAP kinase-interacting kinase 1 (Mnk1) and Mnk2, members of the Ser/Thr protein kinase family, bind tightly to the growth factor-regulated MAP kinases, Erk1 and Erk2. Erk and p38 phosphorylate MNK1 and Mnk2, which stimulates their in vitro kinase activity toward a substrate, eukaryotic initiation factor-4E (eIF-4E). Overexpression of Mnk2 results in increased phosphorylation of endogenous eIF-4E, showing that it can act as an eIF-4E kinase in vivo. Mnk2 may play a role in the response to environmental stress and cytokines. This ubiquitiously expressed protein appears to regulate transcription by phosphorylating eIF-4E, thus increasing the affinity of this protein for the 7-methylguanosine-containing mRNA cap. Expression of active mutants of MNK1 and MNK2 in 293 cells diminishes cap-dependent translation relative to cap-independent translation in a transient reporter assay. Human Mnk2 is homologous to murine Mnk2 (approximately 94% identical) and human Mnk1 (71% identical). In vitro phosphorylation studies show that Mnk2 is a significantly better substrate than Mnk1 for extracellular signal-regulated kinase 2 (Erk2), p38MAPKalpha, and p38MAPKbeta. Mnk2 has also been shown to interact with the C-terminal regions of eIF-4G1 and eIF-4G2.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MKNK2 (MAP kinase interacting serine/threonine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR