Anti-CKBB Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513811
CAPIPA5-13811EA 2286.08 CAD
CAPIPA5-13811
Anti-CKBB Rabbit Polyclonal Antibody
Antibodies
Creatine Kinase (CK) is a dimeric enzyme composed of either M- or B-type subunits. The resulting isoenzymes are expressed at varying levels in different tissues. CK-BB, a cytoplasmic predominantly found in brain tissues, participates in energy homeostasis, reversibly catalyzing the transfer of a phosphate group between ATP and target proteins such as a creatine phosphate. CK-BB exists in normally neglible amounts in the serum of adults, overexpression of CK-BB is associated with cancers of the breast, ovary, prostate, colon, and in small-cell lung cancer. Global assessment of changes in serum levels of CK-BB, CK-MB and CK-MM, are used as a marker for myocardial infarction.

Western Blot: 1:1000

Type: Primary
Antigen: CKBB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR