Anti-Ethanolamine Kinase Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513822
CAPIPA5-13822EA 2561.45 CAD
CAPIPA5-13822
Anti-Ethanolamine Kinase Rabbit Polyclonal Antibody
Ethanolamine kinase 1 (EKI1), functions in the first committed step of the phosphatidylethanolamine synthesis pathway. This cytosolic enzyme is specific for ethanolamine and exhibits negligible kinase activity on choline.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: Ethanolamine Kinase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR