Anti-STK38 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology

PA513831
CAPIPA5-13831EA 2800.97 CAD
CAPIPA5-13831
Anti-STK38 Rabbit Polyclonal Antibody
Antibodies
Stk38,also known as NUCLEAR DBF2-RELATED PROTEIN, contains 1 protein kinase domain that interacts with mob1 and mob2.The homodimeric s100b binds to two molecules of stk38. It is ubiquitously expressed with highest levels observed in peripheral blood leukocytes. Stk38 activated by binding of s100b which releases autoinhibitory n-lobe interactions, enabling atp to bind and the autophosphorylation of ser-281. thr-444 then undergoes calcium- dependent phosphorylation by an upstream kinase. Interactions between phosphorylated thr-444 and the n-lobe promote additional structural changes that complete the activation of the kinase. Autoinhibition is also released by the binding of mob1/mobkl1a and mob2/hcca2 to the n-terminal of stk38.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: STK38 (serine/threonine kinase 38)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR