Anti-CSNK2B Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513834
CAPIPA5-13834EA 2561.45 CAD
CAPIPA5-13834
Anti-CSNK2B Rabbit Polyclonal Antibody
This antibody is predicted to react with bovine, chicken, mouse, porcine, rat, rabbit and Xenopus and zebrafish based on sequence homology. CSNK2B is the beta subunit of casein kinase II, a ubiquitous protein kinase which regulates metabolic pathways, signal transduction, transcription, translation, and replication. The enzyme is composed of three subunits, alpha, alpha prime and beta, which form a tetrameric holoenzyme. The alpha and alpha prime subunits are catalytic, while the beta subunit serves regulatory functions. The enzyme localizes to the endoplasmic reticulum and the Golgi apparatus.

Western Blot: 1:1000

Type: Primary
Antigen: CSNK2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR